Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000-1:5,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Bacteria |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Recombinant Streptomyces sp BenC protein (1-86aa) |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
4.26 mg/mL |
Purification |
Protein G |
Storage buffer |
10mM PBS, pH 7.4, with 50% glycerol |
Contains |
0.03% ProClin 300 |
Storage conditions |
-20°C or -80°C if preferred |
RRID |
AB_2902166 |
This antibody has been tested in direct-ELISA
Carrier of the growing fatty acid chain in fatty acid biosynthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BenC protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support