Substance P (SP) is currently thought to be one of the most important neurotransmitters and neurotransmitters in the brain. It is an undecapeptide belonging to the tachykinin family, which consists of excitatory neuropeptides synthesised in neuronal and glial cells of the human central and peripheral nervous system. Substance P, Neurokinin A, Neuropeptide K, and Neuropeptide gamma are all generated by post-translation cleavage of the precursor Protachykinin-1. Substance P forms the major endogenous ligand for Neurokinin 1 Receptor. The pharmacology of Substance P has been associated with a number of neurological and psychiatric disorders, namely nociception, migraine, asthma, nausea, inflammatory bowel syndrome, urinary incontinence, anxiety and depression. It has also been linked to obesity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-terminal-flanking peptide; MGC26719; neurokinin 1; neurokinin 2; Neurokinin A; neurokinin alpha; Neuromedin L; Neuropeptide gamma; Neuropeptide K; NKA; NPK; PPT; precursor 1; preprotachykinin; preprotachykinin A; protachykinin; Protachykinin-1; RAT TAC; Substance K; Substance P; Tachykinin (substance P, neurokinin A, neuropeptide K, neuropeptide gamma); tachykinin 1; tachykinin 2; tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma)
Gene Aliases: 4930528L02Rik; Hs.2563; NK-1; NK1; NK2; NKA; NKNA; NPK; PPT-A; Ppt5fl; PPTA; PPTA3; RATPPTA3; SP; TAC; TAC1; TAC2
UniProt ID: (Human) P20366, (Rat) P06767, (Mouse) P41539
Entrez Gene ID: (Human) 6863, (Rat) 24806, (Mouse) 21333
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support