Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
CADM4 is involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity. Fukuhara H., Oncogene 20:5401-5407(2001). Grimwood J., Nature 428:529-535(2004)0. Williams Y.N., Oncogene 25:1446-1453(2006).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell adhesion molecule 4; IgSF4C; Immunoglobulin superfamily member 4C; immunoglobulin superfamily, member 4C; membrane glycoprotein Tsll2; NECL-4; nectin-like 4; Nectin-like protein 4; SynCAM 4; TSLC1-like 2; TSLC1-like protein 2; urokinase plasminogen activator receptor
Gene Aliases: CADM4; Igdf4c; IGSF4C; Necl-4; NECL4; synCAM4; TSLL2; Upar
UniProt ID: (Human) Q8NFZ8, (Mouse) Q8R464, (Rat) Q1WIM1
Entrez Gene ID: (Human) 199731, (Mouse) 260299, (Rat) 365216
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support