Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For maximum recovery of product, centrifuge the vial prior to removing the cap.
PRRT1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dispanin subfamily D member 1; DSPD1; interferon induced transmembrane protein domain containing 7; Proline-rich transmembrane protein 1; prrt1 {ECO:0000250|UniProtKB:Q99946}; Synapse differentiation-induced protein 4; SynDIG4
Gene Aliases: Bat-4; Bat4; C6orf31; D17H6S54E; DSPD1; G5b; IFITMD7; NG5; ORF31; PRRT1
UniProt ID: (Human) Q99946, (Rat) Q6MG82, (Mouse) O35449
Entrez Gene ID: (Human) 80863, (Rat) 406167, (Mouse) 260297
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support