Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
FIGURE: 1 / 8
Immunogen sequence: ELQLSVDCAN KTESDLSIEV EFEYPFRLHQ VYFDAPTCRG GTTKVFLVGD YSSSAEFFVT VAVFAFLYSM GALATYIFLQ NKYRENNKGP MLDFLATAVF AFMWLVSSSA WAKGLSDVKM ATDPENIIKE MPVCRQTGNT CKELRDLVTS GLNTSVVFGF LNLVLWVGNL WFVFKETGWA APFLRAPPGA PEKQPAPGDA YGDAGYGQGP GGYGPQDSYG PQGGYQPDYG QPAGSGGSGY GPQGDYGQQG YGPQGAPTSF SNQM
Synaptophysin is a calcium-binding and integral membrane glycoprotein present in presynaptic vesicles in almost all neurons. Synaptophysin has four transmembrane domains and it forms a complex with dynamin at high calcium concentrations suggesting an involvement in synaptic vesicle endocytosis. It is also involved in the regulation of short-term and long-term synaptic plasticity. Synaptophysin is currently the most widely used marker for nerve terminals and for differentiating neuroendocrine tumors. Mutations in the gene can result in mental retardation, X-linked 96.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BM89 antigen; Major synaptic vesicle protein p38; Synaptophysin; Syp I
Gene Aliases: A230093K24Rik; AI848995; MRX96; MRXSYP; p38; Syn; SYP; Syp1
UniProt ID: (Human) P08247, (Rat) P07825, (Mouse) Q62277
Entrez Gene ID: (Human) 6855, (Rat) 24804, (Mouse) 20977
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support