Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
1 µg/mL of MA5-45588 was sufficient for detection of Synaptotagmin-7 in 20 µg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. |Detects approximately 45kDa. Does not cross-react with Synaptotagmin-6 (or others). Can identify other isoforms bands at approximately 65kD.
This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. A similar protein in rodents mediates hormone secretion and lysosome exocytosis. In humans, expression of this gene has been associated with prostate cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IPCA-7; Prostate cancer-associated protein 7; Protein Syt7; synaptotagmin 7; Synaptotagmin VII; Synaptotagmin-7; SYT-7; SytVII
Gene Aliases: AI851541; B230112P13Rik; IPCA-7; IPCA7; PCANAP7; SYT-VII; SYT7; SYTVII
UniProt ID: (Human) O43581, (Mouse) Q9R0N7, (Rat) Q62747
Entrez Gene ID: (Human) 9066, (Mouse) 54525, (Rat) 59267
Molecular Function: membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support