Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSLEDPFFV VRGEVQKAVN TARGLYQRWC ELLQESAAVG REELDWTTNE LRNGLRSIEW DLEDLEETIG IVEANPGKPA AQKSPSDLLD ASAVSATSRY IEEQQATQQL IMDEQDQQLE MVSGSIQVLK HMSGRVGEEL DEQGIMLDAF AQEMDHTQSR MDGVLRKLAK VSHMTSDRRQ WCAIAVLVGV LLLVLILLFS L (1-200 aa encoded by BC017237)
SNARE involved in vesicular transport from the late endosomes to the trans-Golgi network.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Syn10; Syntaxin-10
Gene Aliases: hsyn10; STX10; SYN10
UniProt ID: (Human) O60499
Entrez Gene ID: (Human) 8677
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support