Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total TAAR9.
TAAR9 is a member of a large family of rhodopsin G protein-coupled receptors (GPCRs, or GPRs). GPCRs contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mTaar9; SNORF1 receptor; TaR-3; TaR-9; Trace amine receptor 3; trace amine receptor 9; Trace amine-associated receptor 9; trace-amine-associated receptor 9
Gene Aliases: Gm231; TA3; TAAR9; taR-9; TAR3; TAR9; TRAR3
UniProt ID: (Human) Q96RI9, (Rat) Q923Y6, (Mouse) Q5QD04
Entrez Gene ID: (Human) 134860, (Rat) 319107, (Mouse) 503558
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support