Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: QPNLPSPGTL APTTLQGVVK FQDVSFAYPN RPDRPVLKGL TFTLRPGEVT ALVGPNGSGK STVAALLQNL YQPTGGQVLL DEKPISQYEH CYLHSQVVSV GQEPVLFSGS VRNNIAYGLQ SCEDDKVMAA AQAAHADDFI QEMEHGIYTD VGEKGSQLAA GQKQRLAIAR ALVRDPRVLI LDEATSALDV QCEQALQDWN SRGDRTVLVI AHRLQTVQRA HQILVLQEGK LQKLAQL
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. This gene is located 7 kb telomeric to gene family member ABCB2. The protein encoded by this gene is involved in antigen presentation. This protein forms a heterodimer with ABCB2 in order to transport peptides from the cytoplasm to the endoplasmic reticulum. Mutations in this gene may be associated with ankylosing spondylitis, insulin-dependent diabetes mellitus, and celiac disease. Alternative splicing of this gene produces two products which differ in peptide selectivity and level of restoration of surface expression of MHC class I molecules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABC transporter, MHC 2; Antigen peptide transporter 2; APT2; ATP-binding cassette sub-family B member 3; ATP-binding cassette, sub-family B (MDR/TAP), member 3; Peptide supply factor 2; Peptide transporter involved in antigen processing 2; Peptide transporter PSF2; Peptide transporter TAP2; PSF-2; Really interesting new gene 11 protein; RING11; transporter 2, ABC (ATP binding cassette); transporter 2, ATP-binding cassette, sub-family B (MDR/TAP)
Gene Aliases: ABC18; ABCB3; APT2; D6S217E; PSF-2; PSF2; RING11; TAP2; Y1
UniProt ID: (Human) Q03519
Entrez Gene ID: (Human) 6891
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support