Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CACNG2/4/8 Antibodies

          Antibodies Inc

          TARPGamma2/4/8 Monoclonal Antibody (N245/36)

          View all (10) CACNG2/4/8 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite TARPGamma2/4/8 Monoclonal Antibody (N245/36)

          • Antibody Testing Data (2)
          TARPGamma2/4/8 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TARPGamma2/4/8 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TARPGamma2/4/8 Antibody (75-252) in WB

          Transfected cell immunoblot: extracts of COS cells transiently transfected with GFP- tagged TARPGamma2/3/4/8 or untagged Kv2.1 plasmid and probed with N245/36 (left) and N86/8 (right) TC supe. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TARPGamma2/4/8 Antibody in Western Blot (WB)
          TARPGamma2/4/8 Antibody in Western Blot (WB)

          Product Details

          75-252

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N245/36

          Immunogen

          Fusion protein amino acids 203-323 (cytoplasmic C-terminus) of rat TARPGamma2 (accession number Q71RJ2) produced recombinantly in E. Coli.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4

          Contains

          0.065% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          Product Specific Information

          For maximum recovery of product, centrifuge the vial prior to removing the cap.

          Target Information

          Voltage-dependent calcium channel gamma-2 subunit, TARPGamma2 or Stargazin is encoded by the gene CACNG2 and is the transmembrane gamma subunit of the L-type calcium channel. Stargazin is in involved in regulation and trafficking of the ligand gated ion channel AMPA receptors. It is thought to play a role in learning and memory. Diseases associated with this gene include forms of deafness, Mental Retardation and childhood absnce epilepsy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Neuronal voltage-gated calcium channel gamma-2 subunit; Neuronal voltage-gated calcium channel gamma-4 subunit; Neuronal voltage-gated calcium channel gamma-8 subunit; Stargazin; TARP gamma 2; TARP gamma 8; TARP gamma-2; TARP gamma-4; TARP gamma-8; Transmembrane AMPAR regulatory protein gamma-2; Transmembrane AMPAR regulatory protein gamma-4; Transmembrane AMPAR regulatory protein gamma-8; Voltage-dependent calcium channel gamma-2 subunit; Voltage-dependent calcium channel gamma-4 subunit; Voltage-dependent calcium channel gamma-8 subunit

          View more View less

          Gene Aliases: AI413107; AW060990; AW491861; B230105C07Rik; B930041E13Rik; Cacng2; Cacng4; Cacng8; Ipr328; mir-935; stargazer; stargazin; Stg; wag; waggler

          View more View less

          UniProt ID: (Rat) Q71RJ2, (Mouse) O88602, (Rat) Q8VHW9, (Mouse) Q9JJV4, (Rat) Q8VHW5

          View more View less

          Entrez Gene ID: (Rat) 84347, (Mouse) 12300, (Rat) 140725, (Mouse) 54377, (Rat) 140729, (Mouse) 81905

          View more View less

          Function(s)
          voltage-gated calcium channel activity calcium channel regulator activity protein binding ionotropic glutamate receptor binding voltage-gated ion channel activity calcium channel activity voltage-gated ion channel
          Process(es)
          neuromuscular junction development transmission of nerve impulse regulation of ion transmembrane transport regulation of membrane potential neurological system process membrane depolarization membrane hyperpolarization calcium ion transmembrane transport regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity transport ion transport calcium ion transport
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-56756dfb8b-nml4b:80/100.66.75.22:80.
          git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.1-2025.06.25-1.0