Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Stargazin is a member of the transmembrane AMPAR regulatory proteins (TARP) family and is involved in glutamate receptor trafficking. It has been recently demonstrated that the interaction between stargazin and AMPA receptors is critical for the correct localization of the receptors at the synapse. Phosphorylation of the stargazin protein at Thr 321 by Protein Kinase A regulates its interaction with PSD-95 and synaptic targeting of AMPA receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CaV gamma 2; MGC123981; MGC138502; MGC138504; Neuronal voltage-gated calcium channel gamma-2 subunit; Stargazin; TARP; TARP gamma 2; TARP gamma-2; Transmembrane AMPAR regulatory protein gamma-2; Voltage-dependent calcium channel gamma-2 subunit
Gene Aliases: AW060990; B230105C07Rik; B930041E13Rik; Cacng2; Ipr328; stargazer; stargazin; Stg; wag; waggler
UniProt ID: (Rat) Q71RJ2, (Mouse) O88602
Entrez Gene ID: (Rat) 84347, (Mouse) 12300
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support