Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TCF7 Antibodies

          Proteintech

          TCF7 Recombinant Rabbit Monoclonal Antibody (241154D2)

          View all (20) TCF7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite TCF7 Recombinant Rabbit Monoclonal Antibody (241154D2)

          • Antibody Testing Data (1)
          TCF7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TCF7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TCF7 Antibody (84089-4-RR) in WB

          Various lysates were subjected to SDS PAGE followed by western blot with 84089-4-RR (TCF7 antibody) at dilution of 1:10,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TCF7 Antibody in Western Blot (WB)

          Product Details

          84089-4-RR

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:50,000
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293F

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          241154D2

          Immunogen

          TCF1/TCF7 fusion protein Ag5792

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MPQLDSGGG GAGGGDDLGA PDELLAFQDE GEEQDDKSRD SAAGPERDLA ELKSSLVNES EGAAGGAGIP GVPGAGAGAR GEAEALGREH AAQRLFPDKL PEPLEDGLKA PECTSGMYKE TVYSAFNLLM HYPPPSGAGQ HPQPQPPLHK ANQPPHGVPQ LSLYEHFNSP HPTPAPADIS QKQVHRPLQT PDLSGFYSLT SGSMGQLPHT VSWFTHPSLM LGSGVPGHPA AIPHPAIVPP SGKQELQPFD RNLKTQAESK AEKEAKKPTI KKPLNAFMLY MKEMRAKVIA ECTLKESAAI NQILGRRWHA LSREEQAKYY ELARKERQLH MQLYPGWSAR DNYGKKKRRS REKHQESTTG GKRNAFGTYP EKAAAPAPFL PMTVL

          Target Information

          TCF7 (transcriptional factor 7) is a transcriptional activator involved in T-cell lymphocyte differentiation. Necessary for the survival of CD4+ and CD8+ immature thymocytes. Isoforms for TCF7 lacking the N-terminal CTNNB1 binding domain cannot fulfill this role. TCF7 binds to the T-lymphocyte-specific enhancer element (5'-WWCAAAG-3') found in the promoter of the CD3E gene. It may also act as feedback transcriptional repressor of CTNNB1 and TCF7L2 target genes. TLE1, TLE2, TLE3, and TLE4 repress transactivation mediated by TCF7 and CTNNB1.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: HMG-box; OTTHUMP00000159390; OTTHUMP00000159391; T cell factor-1; T-cell factor 1; T-cell specific; T-cell-specific transcription factor 1; TCF-7; Transcription factor 7; transcription factor 7, T-cell specific

          View more View less

          Gene Aliases: AI465550; TCF-1; TCF1; TCF7

          View more View less

          UniProt ID: (Human) P36402, (Mouse) Q00417

          View more View less

          Entrez Gene ID: (Human) 6932, (Mouse) 21414

          View more View less

          Function(s)
          core promoter binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding protein binding beta-catenin binding sequence-specific DNA binding transcription regulatory region DNA binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding DNA-binding transcription factor gene-specific transcriptional regulator
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter transcription, DNA-templated regulation of transcription from RNA polymerase II promoter immune response neural tube development embryonic genitalia morphogenesis T cell receptor V(D)J recombination regulation of cell proliferation canonical Wnt signaling pathway involved in negative regulation of apoptotic process alpha-beta T cell differentiation embryonic digestive tract morphogenesis embryonic hindgut morphogenesis canonical Wnt signaling pathway cellular response to interleukin-4 beta-catenin-TCF complex assembly regulation of transcription, DNA-templated transcription from RNA polymerase II promoter Wnt signaling pathway generation of neurons
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline