Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | - |
Immunocytochemistry (ICC/IF) |
1:100 | - |
Immunoprecipitation (IP) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Bovine, Dog, Hamster, Human, Mouse, Sheep, Rabbit, Rat |
Host/Isotype |
Rat / IgG2c |
Class |
Monoclonal |
Type |
Antibody |
Clone |
23c |
Immunogen |
Purified recombinant mouse TCP1 alpha construct encoding the C-terminal half of the 1.8 kb full-length Tcp 1b gene expressed in E. coli. Detects the COOH group. |
Conjugate |
APC
|
Excitation/Emission Max |
651/660 nm View spectra |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein G |
Storage buffer |
PBS, pH 7.4, with 50% glycerol |
Contains |
0.1% sodium azide |
Storage conditions |
4° C |
The T Complex Polypeptide 1 (TCP-1) is approximately 60 kDa protein constitutively expressed in almost all eukaryotic cells, and is upregulated during spermatogenesis. It is found in the cytosol as a subunit of a hetero-oligomeric chaperone that is known to be involved in the folding of actin and tubulin. The family of proteins termed chaperonins act to recognize and stabilize polypeptide intermediates during folding, assembly and disassembly, and share many characteristics with Heat Shock Protein 70 (HSP70) including high abundance, induction by environmental stress, and ATPase activity. The chaperonin family includes the mitochondrial HSP60, Escherichia coli GroEL, the plastid Rubisco-subunit binding protein, and the archaebacterial protein TF55. The TCP-1 sequence shows nearly 40% identity to TF55, but only minimal similarity to HSP60 and GroEL.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 65 kDa antigen; CCT-alpha; chaperonin-containing T-complex polypeptide alpha subunit; cytosolic chaperonin containing t-complex polypeptide 1; t-complex polypeptide 1; T-complex protein 1 alpha subunit-like protein; T-complex protein 1 subunit alpha; T-complex protein 1 subunit alpha A; T-complex protein 1 subunit alpha B; T-complex protein 1, alpha subunit; tailless complex polypeptide 1; Tailless complex polypeptide 1A; Tailless complex polypeptide 1B; TCP-1-A; TCP-1-alpha; TCP-1-B
Gene Aliases: AI528772; c-cpn; CCT; CCT-alpha; CCT1; CCTA; CCTalpha; D6S230E; p63; Tcp-1; TCP-1-alpha; TCP1; Tp63; TRiC
UniProt ID: (Bovine) Q32L40, (Human) P17987, (Rat) P28480, (Mouse) P11983, (Chinese hamster) P18279
Entrez Gene ID: (Rabbit) 100302435, (Dog) 484064, (Bovine) 512043, (Sheep) 100135437, (Human) 6950, (Rat) 24818, (Mouse) 21454, (Chinese hamster) 100689476
Molecular Function:
chaperonin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support