Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TCR beta Antibodies

          Invitrogen

          TCR beta Monoclonal Antibody (H57-597), APC, eBioscience™

          35 Published Figures
          91 References
          View all (50) TCR beta antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite TCR beta Monoclonal Antibody (H57-597), APC, eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (35)
          TCR beta Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          TCR beta Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 36

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TCR beta Antibody (17-5961-82) in Flow

          Staining of C57BL/6 splenocytes with staining buffer (autofluorescence) (open histogram) or 0.06 µg of Anti-Mouse TCR beta APC (filled histogram). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)
          TCR beta Antibody in Flow Cytometry (Flow)

          Product Details

          17-5961-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          0.125 µg/test
          View 89 publications 89 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Armenian hamster / IgG

          Recommended Isotype Control

          Armenian Hamster IgG Isotype Control (eBio299Arm), APC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          H57-597

          Conjugate

          APC APC APC
          • Unconjugated
          • Alexa Fluor 532 
          • Alexa Fluor 700 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 496
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant Violet 421
          • eFluor 450
          • eFluor 506
          • FITC
          • Functional Grade
          • NovaFluor Blue 510
          • NovaFluor Blue 530
          • NovaFluor Blue 610-70S
          • NovaFluor Blue 660-120S
          • NovaFluor Blue 725
          • NovaFluor Blue 800
          • NovaFluor UV 765
          • NovaFluor Violet 690
          • NovaFluor Violet 745
          • NovaFluor Violet 800
          • NovaFluor Yellow 810
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          651/660 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_469481

          Product Specific Information

          Description: The H57-597 monoclonal antibody reacts with the beta chain of mouse TCR. TCR beta is expressed by thymocytes in a developmentally regulated manner and a majority of peripheral T cells. Crosslinking of the TCR complex with H57-597 induces activation and proliferation of T cells or apoptosis based on assay conditions. H57-597 is used as a phenotypic marker for TCR beta expressing T cells.

          Applications Reported: The H57-597 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: The H57-597 antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: bTCR; FLJ22602; MGC117436; MGC22624; MGC23964; MGC71411; t-cell antigen receptor; T-cell receptor; TCR beta; Tcra; TCRbeta; TCRD; TRA

          View more View less

          Gene Aliases: TCRbeta; Tib

          View more View less

          Entrez Gene ID: (Mouse) 21577

          View more View less

          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-56756dfb8b-g86sr:80/100.66.72.8:80.
          git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.1-2025.06.25-1.0