The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine
Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. GPSN2 catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA. Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA (Moon and Horton, 2003 [PubMed 12482854]).[supplied by OMIM].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glycoprotein, synaptic 2; Synaptic glycoprotein SC2; TER; Trans-2,3-enoyl-CoA reductase; Very-long-chain enoyl-CoA reductase
Gene Aliases: 2410016D23Rik; A230102P12Rik; AI173355; D17Ertd178e; GPSN2; MRT14; SC2; TECR; TER
UniProt ID: (Human) Q9NZ01, (Mouse) Q9CY27
Entrez Gene ID: (Human) 9524, (Mouse) 106529
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support