Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The target sequence has 85% sequence homology with mouse and rat proteins.
A novel gene designated TERE1, has a significant effect on the growth regulation in bladder cancer. The TERE1 gene maps to a chromosome locus that has been identified by loss of heterozygosity studies as a site of a putative tumor suppressor gene or genes for multiple tumor types including prostate carcinoma. Data suggest that TERE1 may be significant in prostate cancer growth regulation and the down regulation or absence of TERE1 may be an important component of the phenotype of advanced disease. Data also suggest a potential role for this gene product in the progression of bladder cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RP4-796F18.1; transitional epithelia response protein; Transitional epithelial response protein 1; UbiA prenyltransferase domain-containing protein 1
Gene Aliases: SCCD; TERE1; UBIAD1
UniProt ID: (Human) Q9Y5Z9
Entrez Gene ID: (Human) 29914
Molecular Function: transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support