Search Thermo Fisher Scientific
This antibody is predicted to react with human, mouse, dog, pig and cow based on sequence homology.
The protein encoded by this gene belongs to the transglutaminase superfamily. It catalyzes the cross-linking of proteins and the conjugation of polyamines to proteins. Mutations in this gene are associated with spinocerebellar ataxia type 35 (SCA35). Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Protein Aliases: A030011P16; protein-glutamine gamma-glutamyltransferase 6; RP23-20A6.2; transglutaminase 6