Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AKTAKQSAE SVSKGGEKLG RTAAFRALSQ GVESVKKEID DSVLGQTGPY RRPQRLRKRT EFAGDKFKEE KVFEPNEEAL GVVLHKDSKW YQQWKDFKEN NVVFNRFFEM KMKYDESDNA FIRASRALTD KVTDLLGGLF SKTEMSEVLT EILRVDPAFD KDRFLKQCEN DIIPNVLEAM ISGELDILKD WCYEATYSQL AHPIQQAKAL GLQFHSRILD IDNVDLAMGK MMEQGPVLII TFQAQLVMVV RNPKGEVVEG DPDKVLRMLY VWALCRDQDE LNPYAAWRLL DISASSTEQI L
This gene encodes a peripheral membrane protein associated with the mitochondrial inner membrane translocase, which functions in the import of proteins across the mitochondrial inner membrane and into the mitochondrial matrix. The encoded protein mediates binding of mitochondrial heat shock protein 70 to the translocase of inner mitochondrial membrane 23 (TIM23) complex. Expression of this gene is upregulated in kidney in a mouse model of diabetes. A mutation in this gene is associated with familial oncocytic thyroid carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial import inner membrane translocase subunit TIM44; translocase of inner mitochondrial membrane 44 homolog; translocator of inner mitochondrial membrane 44; translocator of inner mitochondrial membrane 44; DNA segment, Chr 8, ERATO Doi 118, expressed
Gene Aliases: 0710005E20Rik; D8Ertd118e; MIMT44; TIM44; TIMM44
UniProt ID: (Human) O43615, (Mouse) O35857, (Rat) O35094
Entrez Gene ID: (Rabbit) 100353480, (Human) 10469, (Pig) 100521124, (Mouse) 21856, (Rat) 29635
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support