MA5-13685 targets TIMP-1 in IHC (P) and WB applications and shows reactivity with Human samples.
The MA5-13685 immunogen is recombinant human TIMP-1.
TIMP1 (metalloproteinase inhibitor 1) is a metalloproteinase inhibitor that functions by forms one-to-one complexes with target metallproteinases, such as collagenases, and irreversible inactivates them by binding to their catalytic zinc cofactor. TIMP1 acts on MMP1, MMP2, MMP3, MMP7, MMP8, MMP9, MMP10, MMP11, MMP12, MMP14, and MMP16. It also functions as a growth factor that regulates cell differenation, migration, and cell death. TIMP1 activates cellular signaling cascades via CD63 and ITGB1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collagenase inhibitor; EPA; erythroid potentiating activity; Erythroid-potentiating activity; Fibroblast collagenase inhibitor; FLJ90373; Metalloproteinase inhibitor; Metalloproteinase inhibitor 1; MMP inhibitor; RP1-230G1.3; TIMP 1; TIMP-1; Tissue inhibitor of metalloproteinases 1
Gene Aliases: CLGI; EPA; EPO; HCI; TIMP; TIMP-1; TIMP1
UniProt ID: (Human) P01033
Entrez Gene ID: (Human) 7076
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support