Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: MILPDRRSVR ESEDEDEEEE ERRQSASRGC TLQYQHATLQ VFTQFHTTSS EGTDQVVTML GPDWLVEVTD LVSDFMRVGD PRVAHMVDSS TLAGLEPGTT PFKVVSPLTE AVLGETLLTV TEEKVSITQL QAQVVASLAL SLRPSPGSSH TILATTAAQQ TLSFLKQEAL LSLWLSYSDG TTAPLSLYSP RDYGLLVSSL DEHV
Required for normal inner ear hair cell function and hearing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Transmembrane protein 132E
Gene Aliases: DFNB99; TMEM132E
UniProt ID: (Human) Q6IEE7
Entrez Gene ID: (Human) 124842
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support