Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: TVFQGVAGQS LQVSCPYDSM KHWGRRKAWC RQLGEKGPCQ RVVSTHNLWL LSFLRRWNGS TAITDDTLGG TLTITLRNLQ PHDAGLYQCQ SLHGSEADTL RKVLVEVLAD PLDHRDAGDL WFPGESESFE DAHVEHSISR PSQGSHLPSC LSKEPLGRRN PLPTHFHPSP PGLHLSHQDS SSQRPLGCSL AWTEARDTST Q
TREM2 is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OTTHUMP00000039771; soluble TREM 2; soluble TREM2; sTREM 2; sTREM2; TREM; TREM-2; Triggering receptor expressed on monocytes 2; Triggering receptor expressed on myeloid cells; Triggering receptor expressed on myeloid cells 2; triggering receptor expressed on myeloid cells 2a; triggering receptor expressed on myeloid cells 2b; triggering receptor expressed on myeloid cells 2c
Gene Aliases: TREM-2; TREM2; Trem2a; Trem2b; Trem2c
UniProt ID: (Human) Q9NZC2, (Mouse) Q99NH8
Entrez Gene ID: (Human) 54209, (Mouse) 83433
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support