Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TRIM21 Antibodies

          Proteintech

          TRIM21 Recombinant Rabbit Monoclonal Antibody (240491B2), CoraLite® Plus 488

          View all (33) TRIM21 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TRIM21 Recombinant Rabbit Monoclonal Antibody (240491B2), CoraLite® Plus 488

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          TRIM21 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TRIM21 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TRIM21 Antibody (CL488-83530-3) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed A549 cells using CoraLite® Plus 488 TRIM21 antibody (CL488-83530-3, Clone: 240491B2) at dilution of 1:0. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TRIM21 Antibody in Immunocytochemistry (ICC/IF)
          TRIM21 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          TRIM21 Recombinant Rabbit Monoclonal Antibody (240491B2), CoraLite® Plus 488

          Product Details

          CL488-83530-3

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -

          Flow Cytometry (Flow)

          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293F

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          240491B2

          Immunogen

          TRIM21 fusion protein Ag28377
          3D Epitope / Immunogen

          Conjugate

          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation

          Excitation/Emission Max

          493/522 nm

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 0.5% BSA, 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: SRIHAEFVQQ KNFLVEEEQR QLQELEKDER EQLRILGEKE AKLAQQSQAL QELISELDRR CHSSALELLQ EVIIVLERSE SWNLKDLDIT SPELRSVCHV PGLKKMLRTC AVHITLDPDT ANPWLILSED RRQVRLGDTQ QSIPGNEERF DSYPMVLGAQ HFHSGKHYWE VDVTGKEAWD LGVCRDSVRR KGHFLLSSKS GFWTIWLWNK QKYEAGTYPQ TPLHLQVPPC QVGIFLDYEA GMVSFYNITD HGSLIYSFSE CAFTGPLRPF FSPGFNDGGK NTAPLTLCPL NIGSQGSTDY

          Target Information

          This gene encodes a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The encoded protein is part of the RoSSA ribonucleoprotein, which includes a single polypeptide and one of four small RNA molecules. The RoSSA particle localizes to both the cytoplasm and the nucleus. RoSSA interacts with autoantigens in patients with Sjogren syndrome and systemic lupus erythematosus. Alternatively spliced transcript variants for this gene have been described but the full-length nature of only one has been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 52 kDa ribonucleoprotein autoantigen Ro/SS-A; 52 kDa Ro protein; 52-kD Ro/SSA ribonucleoprotein; 52kDa ribonucleoprotein autoantigen SS-A/Ro; RING finger protein 81; RING-type E3 ubiquitin transferase TRIM21; ro(SS-A); Ro/SSA 52kDa; Sicca syndrome antigen A; sjoegren syndrome type A antigen; Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro); SS-A; tripartite motif-containing protein 21

          View more View less

          Gene Aliases: RNF81; Ro/SSA; RO52; SSA; SSA1; TRIM21/Ro52

          View more View less

          UniProt ID: (Human) P19474

          View more View less

          Entrez Gene ID: (Human) 6737

          View more View less

          Function(s)
          transcription coactivator activity ubiquitin-protein transferase activity protein binding zinc ion binding identical protein binding metal ion binding ubiquitin protein ligase activity
          Process(es)
          protein polyubiquitination protein monoubiquitination regulation of gene expression proteasomal protein catabolic process positive regulation of autophagy protein ubiquitination protein destabilization positive regulation of protein binding regulation of type I interferon production response to interferon-gamma stress granule disassembly protein K27-linked ubiquitination negative regulation by host of viral release from host cell innate immune response positive regulation of cell cycle negative regulation of innate immune response positive regulation of transcription, DNA-templated protein autoubiquitination cellular response to chemical stress pyroptosis protein K63-linked ubiquitination protein K48-linked ubiquitination protein K6-linked ubiquitination negative regulation of protein deubiquitination antiviral innate immune response positive regulation of NIK/NF-kappaB signaling
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-br959:80/100.66.76.64:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0