Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Phosphorylates phosphatidylinositol (PI) in the first committed step in the production of the second messenger inositol-1, 4, 5, -trisphosphate (PIP). PI4KB may regulate Golgi disintegration/reorganization during mitosis, possibly via its phosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM37; Mulibrey nanism protein; RING-B-box-coiled-coil protein; RING-type E3 ubiquitin transferase TRIM37; TRAF encompassing factor 3; Tripartite motif-containing protein 37
Gene Aliases: 1110032A10Rik; 2810004E07Rik; AI848587; AU043018; KIAA0898; mKIAA0898; MUL; POB1; TEF3; TRIM37
UniProt ID: (Human) O94972, (Mouse) Q6PCX9
Entrez Gene ID: (Human) 4591, (Mouse) 68729
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support