Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MTSPSSSPVF RLETLDGGQE DGSEADRGKL DFGSGLPPME SQFQGEDRKF APQIRVNLNY RKGTGASQPD PNRFDRDRLF NAVSRGVPED LAGLPEYLSK TSKYLTDSEY TEGSTGKTCL MKAVLNLKDG VNACILPLLQ IDRDSGNPQP LVNAQCTDDY YRGHSALHIA IEKRSLQCVK LLVENGANVH ARACGRFFQK GQGTCFYFGE LPLSLAACTK QWDVVSYLLE NPHQPASLQA TDSQGNTVLH ALVMISDNSA ENIALVTSMY DGLLQAGARL CPTVQLEDIR NLQDLTPLKL AAKEGKIEIF RHILQREFSG LSHLSRKFTE WCYGPVRVSL YDLASVDSCE E
VLR-1 is a calcium-permeable, non-selective cation channel with an outward rectification, that seems to be regulated, at least in part, by growth factors, like IGF1, PDGF and morphogenetic neuropeptide/head activator. VRL-1 does not respond to capsaicin, acid, or moderate heat, but instead is activated by high temperatures, with a threshold of approximately 52 degrees C. Within sensory ganglia, VRL-1 is most prominently expressed by a subset of medium- to large-diameter neurons, making it a candidate receptor for transducing high-threshold heat responses in this class of cells. VRL-1 transcripts are not restricted to the sensory nervous system, indicating that this channel may be activated by stimuli other than heat. VRL-1 is one of the six members of the vanilloid-like TRP-channel family which is now known as TRPV family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Osm-9-like TRP channel 2; OTRPC2; RP23-234K24.7; Transient receptor potential cation channel subfamily V member 2; transient receptor potential cation channel, subfamily V, member 2; TrpV2; Vanilloid receptor-like protein 1; VRL-1
Gene Aliases: TRPV2; VRL; VRL-1; VRL1
UniProt ID: (Human) Q9Y5S1
Entrez Gene ID: (Human) 51393
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support