Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the alpha tubulin protein family. Alpha tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Mutations in this gene are associated with polymicrogyria and optic nerve hypoplasia. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-tubulin 8; Tubulin alpha chain-like 2; Tubulin alpha-8 chain; tubulin, alpha 8
Gene Aliases: TUBA8; TUBAL2
UniProt ID: (Human) Q9NY65
Entrez Gene ID: (Human) 51807
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support