Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-82191 recognizes class III beta-tubulin whixh is reported to be restricted to neuronal tissue. Clone TU-20 has been used to investigate tumors of neuronal origin including neuroblastoma and ganglioneuroma. Class III beta tubulin has not been detected in tumors of non neuronal origin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: class II beta-tubulin isotype; class IIb beta-tubulin; Tubulin beta-2B chain; tubulin, beta 2B class IIb; tubulin, beta polypeptide paralog
Gene Aliases: bA506K6.1; PMGYSA; TUBB2B
UniProt ID: (Human) Q9BVA1
Entrez Gene ID: (Human) 347733
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support