Product References
FMRP, FXR1 protein and Dlg4 mRNA, which are associated with fragile X syndrome, are involved in the ubiquitin-proteasome system.
Scientific reports
Shimizu H,Hohjoh H
Thu Feb 02 00:00:00 UTC 2023
Inhibition of Schwann Cell Pyroptosis Promotes Nerve Regeneration in Peripheral Nerve Injury in Rats.
Mediators of inflammation
Wang J,Lu S,Yuan Y,Huang L,Bian M,Yu J,Zou J,Jiang L,Meng D,Zhang J
Mon May 08 00:00:00 UTC 2023
Generation of an induced pluripotent stem cell (iPSC) line (INNDSUi002-A) from a patient with riboflavin-responsive multiple acyl-CoA dehydrogenase deficiency.
Stem cell research
Zhang C,Wang D,Wang Y,Sun P,Zhao D,Zhao X,Zhao Y,Liu F,Xu J,Yan C
Thu Jun 01 00:00:00 UTC 2023
The role of Eya1 and Eya2 in the taste system of mice from embryonic stage to adulthood.
Frontiers in cell and developmental biology
Zhang T,Xu PX
Sat Nov 04 00:00:00 UTC 2023
Alternate-day fasting for the protection of cognitive impairment in c57BL/6J mice following whole-brain radiotherapy.
Neurochemistry international
Wu Q,Yu M,Wang Z,Ai X,Liu Z,Zeng J,Li C,Yuan L,He J,Lin X,Wan W
Sun Jan 01 00:00:00 UTC 2023
Pyramidal neuron morphogenesis requires a septin network that stabilizes filopodia and suppresses lamellipodia during neurite initiation.
Current biology : CB
Radler MR,Liu X,Peng M,Doyle B,Toyo-Oka K,Spiliotis ET
Mon Feb 06 00:00:00 UTC 2023
Bisphenol-A (BPA) Impairs Hippocampal Neurogenesis via Inhibiting Regulation of the Ubiquitin Proteasomal System.
Molecular neurobiology
Singh SJ,Tandon A,Srivastava T,Singh N,Goyal S,Priya S,Chaturvedi RK
Thu Jun 01 00:00:00 UTC 2023
Establishment of an induced pluripotent stem cell (iPSC) line (INNDSUi001-A) from a healthy female Chinese Han.
Stem cell research
Wang D,Zhang R,Wang Y,Sun P,Ji X,Sun X,Xu J,Zhao D,Yan C,Liu F
Fri Jul 01 00:00:00 UTC 2022
Shouhui Tongbian Capsule ameliorates constipation via gut microbiota-5-HT-intestinal motility axis.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Bai J,Cai Y,Huang Z,Gu Y,Huang N,Sun R,Zhang G,Liu R
Sat Oct 01 00:00:00 UTC 2022
Microtubule affinity regulating kinase 4 promoted activation of the NLRP3 inflammasome-mediated pyroptosis in periodontitis.
Journal of oral microbiology
Wang L,Pu W,Wang C,Lei L,Li H
Fri Apr 29 00:00:00 UTC 2022
Silk fibroin nanofibrous scaffolds incorporated with microRNA-222 loaded chitosan nanoparticles for enhanced neuronal differentiation of neural stem cells.
Carbohydrate polymers
Li Z,Meng Z,Zhao Z
Tue Feb 01 00:00:00 UTC 2022
Newly raised anti-c-Kit antibody visualizes morphology of interstitial cells of Cajal in the developing gut of chicken embryos.
Development, growth & differentiation
Yagasaki R,Shikaya Y,Kawachi T,Inaba M,Takase Y,Takahashi Y
Sat Oct 01 00:00:00 UTC 2022
Ambient NO2 exposure induces migraine in rats: Evidence, mechanisms and interventions.
The Science of the total environment
Ye S,Li S,Ma Y,Wei L,Zeng Y,Hu D,Xiao F
Thu Oct 20 00:00:00 UTC 2022
MicroRNA-138 Regulates Spinal Cord Development by Activating the Shh in Fetal Rats.
Pediatric neurosurgery
Ma Z,Li CY,Wang LJ,Xia Y,Feng CA,Peng YF,Han YB,Fan Y,Ba YC
Tue Feb 14 00:00:00 UTC 2023
Periodontal ligaments enhance neurite outgrowth in trigeminal ganglion neurons through Wnt5a production induced by mechanical stimulation.
American journal of physiology. Cell physiology
Takahashi K,Yoshida T,Wakamori M
Thu Dec 01 00:00:00 UTC 2022
Promoted CD4+ T cell-derived IFN-γ/IL-10 by photobiomodulation therapy modulates neurogenesis to ameliorate cognitive deficits in APP/PS1 and 3xTg-AD mice.
Journal of neuroinflammation
Wu X,Shen Q,Chang H,Li J,Xing D
Mon Oct 10 00:00:00 UTC 2022
Promoted CD4+ T cell-derived IFN-γ/IL-10 by photobiomodulation therapy modulates neurogenesis to ameliorate cognitive deficits in APP/PS1 and 3xTg-AD mice.
Journal of neuroinflammation
Wu X,Shen Q,Chang H,Li J,Xing D
Mon Oct 10 00:00:00 UTC 2022
Promoted CD4+ T cell-derived IFN-γ/IL-10 by photobiomodulation therapy modulates neurogenesis to ameliorate cognitive deficits in APP/PS1 and 3xTg-AD mice.
Journal of neuroinflammation
Wu X,Shen Q,Chang H,Li J,Xing D
Mon Oct 10 00:00:00 UTC 2022
Sevoflurane exposure during the second trimester induces neurotoxicity in offspring rats by hyperactivation of PARP-1.
Psychopharmacology
Wang C,Jiang Q,Zhao P
Thu Sep 01 00:00:00 UTC 2022
VPS28 regulates brain vasculature by controlling neuronal VEGF trafficking through extracellular vesicle secretion.
iScience
Dong X,Jiang D,Wang L,Zhao J,Yu L,Huang Y,Wu X,Zhu Y,Zhao Y,Zhao Q,Zhang G,Li X
Fri Apr 15 00:00:00 UTC 2022
Single-nucleus transcriptomic analysis of human dorsal root ganglion neurons.
eLife
Nguyen MQ,von Buchholtz LJ,Reker AN,Ryba NJ,Davidson S
Fri Nov 26 00:00:00 UTC 2021
The Neural Stem Cell Properties of PKD2L1+ Cerebrospinal Fluid-Contacting Neurons in vitro.
Frontiers in cellular neuroscience
Wang S,He Y,Zhang H,Chen L,Cao L,Yang L,Wang C,Pan Y,Tang Q,Tan W,Dou X,Li Q
Fri Apr 02 00:00:00 UTC 2021
Taurine improves the differentiation of neural stem cells in fetal rats with intrauterine growth restriction via activation of the PKA-CREB-BDNF signaling pathway.
Metabolic brain disease
Fang Q,Liu J,Chen L,Chen Q,Ke J,Zhang J,Liu Y,Fu W
Tue Jun 01 00:00:00 UTC 2021
Depletion of Intracellular Glutamine Pools Triggers Toxoplasma gondii Stage Conversion in Human Glutamatergic Neurons.
Frontiers in cellular and infection microbiology
Bando H,Fukuda Y,Watanabe N,Olawale JT,Kato K
Tue Apr 05 00:00:00 UTC 2022
Comparison of Different Tissue Clearing Methods for Three-Dimensional Reconstruction of Human Brain Cellular Anatomy Using Advanced Imaging Techniques.
Frontiers in neuroanatomy
Scardigli M,Pesce L,Brady N,Mazzamuto G,Gavryusev V,Silvestri L,Hof PR,Destrieux C,Costantini I,Pavone FS
Fri Mar 25 00:00:00 UTC 2022
Vezatin regulates seizures by controlling AMPAR-mediated synaptic activity.
Cell death & disease
Wang Y,Yuan J,Yu X,Liu X,Tan C,Chen Y,Xu T
Tue Oct 12 00:00:00 UTC 2021
Neuroprotective Effects of Omentin-1 Against Cerebral Hypoxia/Reoxygenation Injury via Activating GAS6/Axl Signaling Pathway in Neuroblastoma Cells.
Frontiers in cell and developmental biology
Niu X,Cheng Y,Zhang M,Du L,Wu X,Lu C,Li X,Liu S,Zhao A,Zhang S,Wu Z,Ding B,Shi W,Wang C,Yang Y,Tian Y
Wed Feb 16 00:00:00 UTC 2022
miR-221/222 suppression induced by activation of the cAMP/PKA/CREB1 pathway is required for cAMP-induced bidirectional differentiation of glioma cells.
FEBS letters
Liu Q,Tian R,Yu P,Shu M
Mon Nov 01 00:00:00 UTC 2021
miR-221/222 suppression induced by activation of the cAMP/PKA/CREB1 pathway is required for cAMP-induced bidirectional differentiation of glioma cells.
FEBS letters
Liu Q,Tian R,Yu P,Shu M
Mon Nov 01 00:00:00 UTC 2021
miR-221/222 suppression induced by activation of the cAMP/PKA/CREB1 pathway is required for cAMP-induced bidirectional differentiation of glioma cells.
FEBS letters
Liu Q,Tian R,Yu P,Shu M
Mon Nov 01 00:00:00 UTC 2021
ESRRB Facilitates the Conversion of Trophoblast-Like Stem Cells From Induced Pluripotent Stem Cells by Directly Regulating CDX2.
Frontiers in cell and developmental biology
Yu S,Zhang R,Shen Q,Zhu Z,Zhang J,Wu X,Zhao W,Li N,Yang F,Wei H,Hua J
Fri Oct 08 00:00:00 UTC 2021
Generation of an EFNB2-2A-mCherry reporter human embryonic stem cell line using CRISPR/Cas9-mediated site-specific homologous recombination.
Stem cell research
Huang Y,Wu H,Han X,Wu J,Yu M,Zhao ZA,Shen Z,Hu S,Lei W
Thu Apr 01 00:00:00 UTC 2021
CNTs-CaP/chitosan-coated AZ91D magnesium alloy extract promoted rat dorsal root ganglia neuron growth via activating ERK signalling pathway.
Cell biochemistry and function
Liu T,Li Q,Yang S,Zhao T,Lin J,Ju T,Wen Z
Fri Oct 01 00:00:00 UTC 2021
Posterior Neocortex-Specific Regulation of Neuronal Migration by CEP85L Identifies Maternal Centriole-Dependent Activation of CDK5.
Neuron
Kodani A,Kenny C,Lai A,Gonzalez DM,Stronge E,Sejourne GM,Isacco L,Partlow JN,O'Donnell A,McWalter K,Byrne AB,Barkovich AJ,Yang E,Hill RS,Gawlinski P,Wiszniewski W,Cohen JS,Fatemi SA,Baranano KW,Sahin M,Vossler DG,Yuskaitis CJ,Walsh CA
Wed Apr 22 00:00:00 UTC 2020
Propofol Exposure Disturbs the Differentiation of Rodent Neural Stem Cells via an miR-124-3p/Sp1/Cdkn1b Axis.
Frontiers in cell and developmental biology
Cao J,Li Y,Zeng F,Liu X,Tao T,Qin Z
Thu Oct 01 00:00:00 UTC 2020
Zinc Promotes Patient-Derived Induced Pluripotent Stem Cell Neural Differentiation via ERK-STAT Signaling.
Stem cells and development
Yang M,Bao D,Shi A,Yuan H,Wang J,He W,Tong X,Qin H
Wed Jul 01 00:00:00 UTC 2020
Co-delivery of brinzolamide and miRNA-124 by biodegradable nanoparticles as a strategy for glaucoma therapy.
Drug delivery
Li T,Wang Y,Chen J,Gao X,Pan S,Su Y,Zhou X
Tue Dec 01 00:00:00 UTC 2020
A single factor induces neuronal differentiation to suppress glioma cell growth.
CNS neuroscience & therapeutics
Fu JQ,Chen Z,Hu YJ,Fan ZH,Guo ZX,Liang JY,Ryu BM,Ren JL,Shi XJ,Li J,Jia S,Wang J,Ke XS,Ma X,Tan X,Zhang T,Chen XZ,Zhang C
Mon Apr 01 00:00:00 UTC 2019
A single factor induces neuronal differentiation to suppress glioma cell growth.
CNS neuroscience & therapeutics
Fu JQ,Chen Z,Hu YJ,Fan ZH,Guo ZX,Liang JY,Ryu BM,Ren JL,Shi XJ,Li J,Jia S,Wang J,Ke XS,Ma X,Tan X,Zhang T,Chen XZ,Zhang C
Mon Apr 01 00:00:00 UTC 2019
Chlorogenic acid effectively treats cancers through induction of cancer cell differentiation.
Theranostics
Huang S,Wang LL,Xue NN,Li C,Guo HH,Ren TK,Zhan Y,Li WB,Zhang J,Chen XG,Han YX,Zhang JL,Jiang JD
Fri Jul 24 00:00:00 UTC 2020
Epigenetic suppression of liver X receptor β in anterior cingulate cortex by HDAC5 drives CFA-induced chronic inflammatory pain.
Journal of neuroinflammation
Li YJ,Zhang K,Sun T,Wang J,Guo YY,Yang L,Yang Q,Li YJ,Liu SB,Zhao MG,Wu YM
Sat Jun 29 00:00:00 UTC 2019
Functional analysis of juxta- and intra-membrane domains of murine APP by genome editing in Neuro2a cells.
Biochemical and biophysical research communications
Kaneshiro N,Imaoka R,Komai M,Kashiyama T,Sakurai T,Uehara T,Takasugi N
Mon Jul 02 00:00:00 UTC 2018
Functional analysis of juxta- and intra-membrane domains of murine APP by genome editing in Neuro2a cells.
Biochemical and biophysical research communications
Kaneshiro N,Imaoka R,Komai M,Kashiyama T,Sakurai T,Uehara T,Takasugi N
Mon Jul 02 00:00:00 UTC 2018
Dexmedetomidine Protects Mouse Brain from Ischemia-Reperfusion Injury via Inhibiting Neuronal Autophagy through Up-Regulating HIF-1α.
Frontiers in cellular neuroscience
Luo C,Ouyang MW,Fang YY,Li SJ,Zhou Q,Fan J,Qin ZS,Tao T
Fri Apr 08 00:00:00 UTC 2022
Dexmedetomidine Protects Mouse Brain from Ischemia-Reperfusion Injury via Inhibiting Neuronal Autophagy through Up-Regulating HIF-1α.
Frontiers in cellular neuroscience
Luo C,Ouyang MW,Fang YY,Li SJ,Zhou Q,Fan J,Qin ZS,Tao T
Fri Apr 08 00:00:00 UTC 2022
A Human Long Non-Coding RNA ALT1 Controls the Cell Cycle of Vascular Endothelial Cells Via ACE2 and Cyclin D1 Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Li W,Wang R,Ma JY,Wang M,Cui J,Wu WB,Liu RM,Zhang CX,Li W,Wang SM
Tue Dec 26 00:00:00 UTC 2017
A Human Long Non-Coding RNA ALT1 Controls the Cell Cycle of Vascular Endothelial Cells Via ACE2 and Cyclin D1 Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Li W,Wang R,Ma JY,Wang M,Cui J,Wu WB,Liu RM,Zhang CX,Li W,Wang SM
Tue Dec 26 00:00:00 UTC 2017