Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: AVISHERAQ QQAIALAKEV SCPMSSSQEV VSCLRQKPAN VLNDAQTKLL AVSGPFHYWG PVIDGHFLRE PPARALKRSL RVEVDLLIGS SQDDGLINRA KAVKQFEESQ GRTSSKTAFY QALQNSLGGE DSDARVEAAA TWYYSLEHST DDYASFSRAL ENATRDYFII CPIIDMASAW AKRARGNVFM YHAPENYGHG SLELLADVQF ALGLPFYPAY EGQFSLEEKS LSLKIMQYFS HFIRSGNPNY PYEFSRKVPT FATPWPDFVP RAGGENYKEF SELLPNRQGL KKADCSFWSK YISSLKTSAD GAKGGQSAES EEEELTAGSG LREDLLSLQE PGSKTYSK (2422-2768 aa encoded by BC140933)
The human thyroglobulin (hTg) is a high molecular weight glycoprotein (605 kDa) found in the thyroid follicular cells. It plays a central role in the uptake, incorporation, and regulated biosynthesis of thyroid hormones, T4 and T3. Thyroid disorders are, in large part, due to autoimmune origin, and anti-thyroglobulin autoantibodies were the first factor to be discovered. Anti-hTG is found in all thyroid autoimmune diseases (Hashimoto thyroiditis, Graves diseases), with the highest level observed in Hashimoto thyroiditis. Anti-hTG is also characteristic of thyroid cancer, and its determination can be used for the follow up of cancer patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Colloid; Tg; Thyroglobulin
Gene Aliases: AITD3; TG; TGN
UniProt ID: (Human) P01266
Entrez Gene ID: (Human) 7038
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support