Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
FIGURE: 1 / 3
Immunogen sequence: DKATGYHYR CITCEGCKGF FRRTIQKNLH PSYSCKYEGK CVIDKVTRNQ CQECRFKKCI YVGMATDLVL DDSKRLAKRK LIEENREKRR REELQKSIGH KPEPTDEEWE LIKTVTEAHV ATNAQGSHWK QKRKFLPEDI GQAPIVNAPE GGKVDLEAFS HFTKIITPAI TRVVDFAKKL PMFCELPCED QIILLKGCCM EIMSLRAAVR YDPESETLTL NGEMAVTRGQ LKNGGLGVVS DAIFDLGMSL SSFNLDDTEV ALLQAVLLMS SDRPGLACVE RIEKYQDSFL LAFEHYINYR KHHVTHFWPK LLMKVTDLRM IGACHASRFL HMKVECPTEL FPPLFLEVFE D
Thyroid hormone receptors (TR), also known as c-erbA-2, are ligand-dependent, intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences called hormone response elements following activation by the appropriate hormone. Thyroid hormones, through their interaction with TR, effect metabolic processes, growth and development in many tissues by regulating the expression of genes for growth hormone, malic enzyme and several hepatic proteins. There are two forms of TR; TR alpha (on chromosome 17) and TR beta (on chromosome 3). Each of these isoforms also has two isoforms; TR alpha-1 and TRv alpha-2, and TR beta-1 and TR beta-2 respectively. TR alpha-1 and 2 are identical through amino acid 370 where their sequences diverge. TRv alpha-2, which does not bind T3 and is a strong negative regulator of the functional TR, is 80 amino acids longer than TR alpha-1. The corresponding gene for the thyroid hormone receptor beta-1 is NR1A2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-erbA-2; c-erbA-beta; ERBA 2; ERBA BETA; ERBA-BETA; MGC126109; MGC126110; Nuclear receptor subfamily 1 group A member 2; oncogene ERBA2; thyroid hormone nuclear receptor beta variant 1; Thyroid hormone receptor beta; thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, avian); TR; TR beta; TR beta 1; TR-B1C-ERBA-2
Gene Aliases: C-ERBA-2; C-ERBA-BETA; c-erbAbeta; ERBA2; GRTH; NR1A2; PRTH; T3R[b]; T3Rbeta; THR1; THRB; THRB1; THRB2
UniProt ID: (Human) P10828, (Mouse) P37242
Entrez Gene ID: (Human) 7068, (Mouse) 21834
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support