Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 6
Recommended positive controls: 293T, A431, Jurkat, Raji, NIH-3T3.
Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (99%), Xenopus laevis (99%), Chicken (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha-chain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: class IVb beta tubulin; epididymis secretory sperm binding protein; microtubule protein; novel tubulin protein; tubulin beta; Tubulin beta-2 chain; Tubulin beta-2C chain; Tubulin beta-4B chain; tubulin, beta 2C; tubulin, beta 2c1; tubulin, beta, 2; unnamed protein product
Gene Aliases: 4930542G03Rik; Beta2; LCAEOD; TUBB2; TUBB2C; Tubb2c1; TUBB4B
UniProt ID: (Human) P68371, (Rat) Q6P9T8, (Mouse) P68372
Entrez Gene ID: (Human) 10383, (Rat) 296554, (Mouse) 227613
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support