Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SQDGASQFQE VIRQELELSV KKELEKILTT ASSHEFEHTK KDLDGFRKLF HRFLQEKGPS VDWGKIQRPP EDSIQPYEKI KARGLPDNIS SVLNKLVVVK LNGGLGTSMG CKGPKSLIGV RNENTFLDLT VQQIEHLNKT YNTDVPLVLM NSFNTDEDTK KILQKYNHCR VKIYTFNQSR YPRINKESLL PVAKDVSYSG ENTEAWYPPG HGDIYASFYN SGLLDTFIGE GKEYIFVSNI DNLGATVDLY ILNHLMNPPN GKRCEFVMEV TNKTRADVKG GTLTQY
The enzyme encoded by this gene is an important intermediary in mammalian carbohydrate interconversions. It transfers a glucose moiety from glucose-1-phosphate to MgUTP and forms UDP-glucose and MgPPi. In liver and muscle tissue, UDP-glucose is a direct precursor of glycogen; in lactating mammary gland it is converted to UDP-galactose which is then converted to lactose. The eukaryotic enzyme has no significant sequence similarity to the prokaryotic enzyme. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: testis tissue sperm-binding protein Li 58p; UDP glucose pyrophosphorylase; UDP-glucose diphosphorylase; UDP-glucose pyrophosphorylase; UDP-glucose pyrophosphorylase 1; UDPGP; UGPase; UGPase 2; uridindiphosphoglucosepyrophosphorylase 2; uridyl diphosphate glucose pyrophosphorylase 2; Uridyl diphosphate glucose pyrophosphorylase-1; UTP--glucose-1-phosphate uridylyltransferase; UTP--glucose-1-phosphate uridylyltransferase 2; UTP-glucose-1-phosphate uridyltransferase
Gene Aliases: pHC379; UDPG; UDPGP; UDPGP2; UGP1; UGP2; UGPase; UGPP1; UGPP2
UniProt ID: (Human) Q16851, (Pig) P79303, (Mouse) Q91ZJ5
Entrez Gene ID: (Human) 7360, (Rabbit) 100350632, (Pig) 397040, (Rat) 289827, (Mouse) 216558
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support