Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: IEAVGGKLS VTATRENMAY TVECLRGDVD ILMEFLLNVT TAPEFRRWEV ADLQPQLKID KAVAFQNPQT HVIENLHAAA YRNALANPLY CPDYRIGKVT SEELHYFVQN HFTSARMALI GLGVSHPVLK QVAEQFLNMR GGLGLSGAKA NYRGGEIREQ NGDSLVHAAF VAESAVAGSA EANAFSVLQH VLGAGPHVKR GSNTTSHLHQ AVAKATQQPF DVSAFNASYS DSGLFGIYTI SQATAAGDVI KAAYNQVKTI AQGNLSNTDV QAAKNKLKAG YLMSVESSEC FLEEVGSQAL VAGSYMPPST VLQQIDSVAN ADIINAAKKF VSGQKSMAAS GNLGHTPFVD EL
This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. The core protein 2 is required for the assembly of the complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complex III subunit 2; Core protein II; Cytochrome b-c1 complex subunit 2, mitochondrial; cytochrome bc-1 complex core protein II; U MC3DN5; Ubiquinol-cytochrome-c reductase complex core protein 2
Gene Aliases: MC3DN5; QCR2; UQCR2; UQCRC2
UniProt ID: (Human) P22695
Entrez Gene ID: (Human) 7385
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support