The antibody is specific to USP50.
Has no peptidase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 50; Inactive ubiquitin carboxyl-terminal hydrolase 50; Inactive ubiquitin-specific peptidase 50; Putative ubiquitin carboxyl-terminal hydrolase 50; ubiquitin specific peptidase 50; ubiquitin specific protease 50; Ubiquitin thioesterase 50; ubiquitin thiolesterase 50; Ubiquitin-specific-processing protease 50
Gene Aliases: 1700086G18Rik; 4930511O11Rik; RGD1564751; USP50
UniProt ID: (Human) Q70EL3, (Mouse) Q6P8X6
Entrez Gene ID: (Human) 373509, (Mouse) 75083, (Rat) 311399
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support