Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • VAPA/VAPB Antibodies

          Antibodies Inc

          VAPA/B Monoclonal Antibody (N479/107), FL550

          View all (7) VAPA/VAPB antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite VAPA/B Monoclonal Antibody (N479/107), FL550

          Product Details

          75-496-FL550

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N479/107

          Immunogen

          Fusion protein amino acids 1-219 (N-terminus) of rat VAPA (accession number Q9Z270) produced recombinantly in E. Coli.
          View immunogen

          Conjugate

          FL550 FL550 FL550
          • Unconjugated
          • FL490
          • FL594
          • FL650
          • Request custom conjugation

          Excitation/Emission Max

          550/575 nm

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          Product Specific Information

          For maximum recovery of product, centrifuge the vial prior to removing the cap.

          Target Information

          VAPA/VAPB may play a role in vesicle trafficking. Present in the plasma membrane and in intracellular vesicles. Interacts with VAMP1 and VAMP2. Subunit: Homodimer, and heterodimer with VAPA. Subcellular Location: Cell membrane; Single-pass type IV membrane protein. Intracytoplasmic membrane; Single-pass type IV membrane protein. Tissue Specificity: Ubiquitous. Isoform 1 predominates. Disease: Defects in VAPB are the cause of amyotrophic lateral sclerosis type 8 (ALS8). ALS8 is a familial form of amyotrophic lateral sclerosis, a neurodegenerative disorder affecting upper and lower motor neurons and resulting in fatal paralysis. Sensory abnormalities are absent. Death usually occurs within 2 to 5 years. The etiology of amyotrophic lateral sclerosis is likely to be multifactorial, involving both genetic and environmental factors. The disease is inherited in 5-10% of cases leading to familial forms. Defects in VAPB are a cause of proximal adult autosomal dominant spinal muscular atrophy; also called late-onset spinal muscular atrophy Finkel type. Spinal muscular atrophies are neurodegenerative disorders characterized by degeneration of lower motor neurons, leading to progressive paralysis muscular atrophy. This form is a late- adult-onset form of the disease (after age 20 years). The patients show a benign course, most of them remaining ambulatory 10 to 40 years after clinical onset.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 33 kDa VAMP-associated protein; synaptobrevin-associated protein of 33kDa; VAMP-A; VAMP-associated protein 33b; VAMP-associated protein A; VAMP-associated protein B; VAMP-B; VAP-33; VAP-A; VAP-B; vesicle-associated membrane protein, associated protein a; vesicle-associated membrane protein, associated protein B and C; Vesicle-associated membrane protein-associated protein A; Vesicle-associated membrane protein-associated protein B

          View more View less

          Gene Aliases: 33kDa; AI225786; AI840687; AI848259; D2Abb2e; R75548; Vamp33b; VAP33; VAP33b; Vapa; Vapb

          View more View less

          UniProt ID: (Rat) Q9Z270, (Mouse) Q9WV55, (Rat) Q9Z269

          View more View less

          Entrez Gene ID: (Rat) 58857, (Mouse) 30960, (Rat) 60431, (Mouse) 56491

          View more View less

          Function(s)
          signal transducer activity microtubule binding protein domain specific binding FFAT motif binding protein heterodimerization activity molecular_function enzyme binding protein homodimerization activity beta-tubulin binding membrane traffic protein membrane trafficking regulatory protein
          Process(es)
          ER to Golgi vesicle-mediated transport endoplasmic reticulum organization signal transduction cell death neuron projection development protein folding in endoplasmic reticulum positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation by host of viral release from host cell negative regulation by host of viral genome replication positive regulation by host of viral genome replication protein localization to endoplasmic reticulum COPII-coated vesicle budding cellular calcium ion homeostasis activation of signaling protein activity involved in unfolded protein response biological_process modulation by virus of host morphology or physiology endoplasmic reticulum unfolded protein response negative regulation by host of viral release from host cell positive regulation of viral genome replication negative regulation by virus of viral protein levels in host cell response to unfolded protein
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6ff95d844f-krbjv:80/100.66.78.119:80.
          git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.28.0-Offline