Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • VE-cadherin (CD144) Antibodies

          Proteintech

          VE-cadherin/CD144 Polyclonal Antibody

          View all (69) VE-cadherin (CD144) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite VE-cadherin/CD144 Polyclonal Antibody

          • Antibody Testing Data (2)
          VE-cadherin/CD144 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          VE-cadherin/CD144 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          VE-cadherin/CD144 Antibody (32332-1-AP) in ICC/IF

          Immunofluorescent analysis of (-20°C Methanol) fixed bEnd.3 cells using VE-cadherin/CD144 antibody (32332-1-AP) at dilution of 1:200 and CoraLite488-Conjugated Goat Anti-Rabbit IgG (H+L) (SA,00013-2). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          VE-cadherin/CD144 Antibody in Immunocytochemistry (ICC/IF)
          VE-cadherin/CD144 Antibody in Western Blot (WB)
          VE-cadherin/CD144 Polyclonal Antibody

          Product Details

          32332-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000-1:10,000
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          VE-cadherin/CD144 fusion protein Eg0921
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          350 µg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Sequence of this protein is as follows: DWIWNQMHID EEKNESLPHY VGKIKSNVNR QNAKYVLQGE FAGKIFGVDA NTGNVLAYER LDREKVSEYF LTALIVDKNT NKNLEQPSSF TVKVHDINDN WPVFSHQVFN ASVPEMSAIG TSVIRVTAVD ADDPTVAGHA TVLYQIVKGN EYFSIDNSGL IFTKIKNLDR EKQAEYKIVV ETQDALGLRG ESGTATVMIR LEDINDNFPV FTQSTYTFSV PEDIRVGKPL GFLTVVDPDE PQNRMTKYSI MQGEYRDTFT IETDPKRNEG IIKPTKSLDY EVIQQYTFYI EATDPTIRYE YLSSTSGKNK AMVTINVLDV DEPPVFQRHF YHFKLPENQK KPLIGTVVAK DPDKAQRSIG YSIRKTSDRG QFFRITKQGN IYNEKELDRE TYAWYNLTVE ANELDSRGNP VGKESIVQVY IEVLDENDNP PEFAQPYEPK VCENAAQGKL VVQISATDKD VVPVNPKFKF ALKNEDSNFT LINNHDNTAN ITVKYGQFNR EHAKFHYLPV LISDNGVPSL TGTSTLTVGV CKCNEQGEFT FCEEMAA

          Target Information

          VE-cadherin is a member of the cadherin superfamily that is located in a six-cadherin cluster in a region on the long arm of chromosome 16 and is involved in loss of heterozygosity events in breast and prostate cancer. VE-cadherin protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Functioning as a classic cadherin by imparting to cells the ability to adhere in a homophilic manner, VE-cadherin may play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. An alternative splice variant has been described but the full length sequence of VE-cadherin has not been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 7B4/cadherin-5; Cadherin-5; CD144; Vascular endothelial cadherin; VE-cadherin

          View more View less

          Gene Aliases: 7B4; AA408225; Cd144; Cdh5; VE-Cad; Vec; VEcad; VECD

          View more View less

          UniProt ID: (Mouse) P55284

          View more View less

          Entrez Gene ID: (Mouse) 12562, (Rat) 307618

          View more View less

          Function(s)
          receptor binding calcium ion binding protein binding beta-catenin binding protein phosphatase binding ion channel binding metal ion binding cadherin
          Process(es)
          blood vessel maturation cell adhesion homophilic cell adhesion via plasma membrane adhesion molecules negative regulation of cell proliferation single organismal cell-cell adhesion regulation of establishment of cell polarity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-57b588f4f5-kdjgd:80/100.66.79.206:80.
          git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.39.0-Offline