Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | - |
Immunohistochemistry (IHC) |
- | View 1 publication 1 publication |
Immunohistochemistry (PFA fixed) (IHC (PFA)) |
1:100 | - |
Immunocytochemistry (ICC/IF) |
1:100 | View 1 publication 1 publication |
Immunocytochemistry (ICC) |
- | View 1 publication 1 publication |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Published species |
Mouse |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
S29-29 |
Immunogen |
Fusion protein amino acids 501-582 (cytoplasmic C-terminus) of rat VGlut2 |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein G |
Storage buffer |
PBS, pH 7.4, with 50% glycerol |
Contains |
0.09% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2735411 |
VGLU2 mediates the uptake of glutamate into synaptic vesicles at presynaptic nerve terminals of excitatory neural cells. This protein may also mediate the transport of inorganic phosphate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: differentation-associated Na-dependent inorganic phosphate cotransporter; Differentiation-associated BNPI; Differentiation-associated Na(+)-dependent inorganic phosphate cotransporter; differentiation-associated Na-dependent inorganic phosphate cotransporter; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6; solute carrier family 17 (vesicular glutamate transporter), member 6; Solute carrier family 17 member 6; Vesicular glutamate transporter 2; VGlU2; VGluT2
Gene Aliases: 2900073D12Rik; DNPI; SLC17A6; VGLUT2
UniProt ID: (Human) Q9P2U8, (Mouse) Q8BLE7, (Rat) Q9JI12
Entrez Gene ID: (Human) 57084, (Mouse) 140919, (Rat) 84487
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support