Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MTAEFLSLLC LGLCLGYEDE KKNEKPPKPS LHAWPSSVVE AESNVTLKCQ AHSQNVTFVL RKVNDSGYKQ EQSSAENEAE FPFTDLKPKD AGRYFCAYKT TASHEWSESS EHLQLVVTDK HDELEAPSMK TDTRTIFVAI FSCISILLLF LSVFIIYRCS QHSELRERKG REGE
VSTM1 (V-Set And Transmembrane Domain Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include cytokine activity. An important paralog of this gene is LILRB1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LAIR homolog; OSCAR-like transcript-1; Signal inhibitory receptor on leukocytes-1; SIRL-1; V-set and transmembrane domain-containing protein 1
Gene Aliases: SIRL-1; SIRL1; UNQ3033; UNQ3033/PRO9835; VSTM1
UniProt ID: (Human) Q6UX27
Entrez Gene ID: (Human) 284415
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support