Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • WNT5A Antibodies

          Invitrogen

          WNT5A Recombinant Rabbit Monoclonal Antibody (K01_2K15)

          View all (24) WNT5A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite WNT5A Recombinant Rabbit Monoclonal Antibody (K01_2K15)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          WNT5A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          WNT5A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          WNT5A Antibody (MA563928) in ICC/IF

          Immunocytochemistry analysis of WNT5A (green) in HeLa cells. Samples were incubated with WNT5A recombinant monoclonal (Product # MA563928). DAPI (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          WNT5A Antibody in Immunocytochemistry (ICC/IF)
          WNT5A Antibody in Western Blot (WB)
          WNT5A Recombinant Rabbit Monoclonal Antibody (K01_2K15)

          Product Details

          MA563928

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293F

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          K01_2K15

          Immunogen

          A synthetic peptide of human Wnt5a (aa 340-354).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A/G

          Storage buffer

          50mM tris glycine, pH 7.4, with 0.05% BSA, 40% glycerol, 150mM NaCl

          Contains

          0.01% sodium azide

          Storage conditions

          Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3704976

          Product Specific Information

          Immunogen sequence: CGRGYDQFKT VQTER

          Target Information

          Wnt5A is a secreted protein found in the extracellular region that belongs to the Wnt family. It is a proto-oncogene that functions in modulating cell fate and behavior during vertebrate development and has an important role in Wnt signaling pathway. It is a glycoprotein that plays an important role in cell communication and development. Wnt5A acts as a ligand for members of the frizzled family of seven transmembrane receptors. It is suspected that Wnt5A is a developmental protein. Wnt5A is a signaling molecule which affects the development of discrete regions of tissues and is likely to signal over only few cell diameters.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: epididymis secretory sperm binding protein; hWNT5A; OTTHUMP00000212837; OTTHUMP00000212861; Protein Wnt-5a; unnamed protein product; wingless-type MMTV integration site family, member 5A; WNT-5A protein

          View more View less

          Gene Aliases: hWNT5A; WNT5A

          View more View less

          UniProt ID: (Human) P41221

          View more View less

          Entrez Gene ID: (Human) 7474

          View more View less

          Function(s)
          receptor binding frizzled binding receptor tyrosine kinase-like orphan receptor binding cytokine activity protein binding phospholipid binding protein domain specific binding receptor agonist activity chemoattractant activity involved in axon guidance
          Process(es)
          establishment of planar polarity somitogenesis positive regulation of cytokine production kidney development epithelial to mesenchymal transition neural tube closure positive regulation of endothelial cell proliferation heart looping morphogenesis of an epithelium positive regulation of mesenchymal cell proliferation lens development in camera-type eye positive regulation of cytokine production involved in immune response primary heart field specification secondary heart field specification ventricular septum development atrial septum development type B pancreatic cell development pericardium morphogenesis axis elongation optic cup formation involved in camera-type eye development inflammatory response phospholipase C-activating G-protein coupled receptor signaling pathway Wnt signaling pathway, calcium modulating pathway JNK cascade determination of left/right symmetry axonogenesis axon guidance hindgut morphogenesis midgut development protein localization cell proliferation fibroblast growth factor receptor signaling pathway male gonad development anterior/posterior axis specification, embryo anterior/posterior pattern specification mesenchymal cell proliferation positive regulation of endothelial cell migration positive regulation of gene expression epithelial cell migration macrophage derived foam cell differentiation positive regulation of T cell chemotaxis positive regulation of neuron projection development Wnt signaling pathway cell migration olfactory bulb interneuron development neural tube development neurogenesis cell differentiation neuron differentiation keratinocyte differentiation lung development embryonic limb morphogenesis BMP signaling pathway negative regulation of BMP signaling pathway midbrain development positive regulation of chemokine production positive regulation of interferon-gamma production positive regulation of interleukin-1 beta production positive regulation of interleukin-6 production positive regulation of interleukin-8 production positive regulation of tumor necrosis factor production regulation of protein localization positive regulation of Rac protein signal transduction limb morphogenesis non-canonical Wnt signaling pathway post-anal tail morphogenesis chemoattraction of serotonergic neuron axon chemorepulsion of dopaminergic neuron axon negative regulation of fibroblast growth factor receptor signaling pathway wound healing inner ear morphogenesis embryonic digit morphogenesis positive regulation of macrophage activation negative regulation of apoptotic process regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of MAPK cascade cell fate commitment establishment of epithelial cell apical/basal polarity negative regulation of fat cell differentiation positive regulation of protein catabolic process positive regulation of angiogenesis positive regulation of ossification positive regulation of endocytosis positive regulation of meiotic nuclear division negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade development of primary male sexual characteristics negative regulation of melanin biosynthetic process positive regulation of fibroblast proliferation paraxial mesoderm formation digestive tract morphogenesis notochord morphogenesis embryonic skeletal system development genitalia development neuron projection morphogenesis negative regulation of axon extension involved in axon guidance axon extension involved in axon guidance hypophysis morphogenesis epithelial cell proliferation positive regulation of epithelial cell proliferation negative regulation of epithelial cell proliferation regulation of inflammatory response positive regulation of inflammatory response regulation of synapse organization negative chemotaxis cartilage development positive regulation of anagen positive regulation of protein kinase B signaling negative regulation of synapse assembly convergent extension convergent extension involved in axis elongation convergent extension involved in organogenesis uterus development cervix development vagina development canonical Wnt signaling pathway Wnt signaling pathway, planar cell polarity pathway urinary bladder development face development positive regulation of type I interferon-mediated signaling pathway lateral sprouting involved in mammary gland duct morphogenesis tube closure mesenchymal-epithelial cell signaling negative regulation of prostatic bud formation mammary gland branching involved in thelarche epithelial cell proliferation involved in mammary gland duct elongation positive regulation of response to cytokine stimulus regulation of branching involved in mammary gland duct morphogenesis mesodermal to mesenchymal transition involved in gastrulation positive regulation of macrophage cytokine production positive regulation of cartilage development somite development secondary palate development thymocyte apoptotic process positive regulation of thymocyte apoptotic process cellular response to molecule of bacterial origin cellular response to lipopolysaccharide cellular response to calcium ion cellular response to retinoic acid cellular response to interferon-gamma hematopoietic stem cell proliferation dopaminergic neuron differentiation cellular response to transforming growth factor beta stimulus stem cell proliferation negative regulation of mesenchymal cell proliferation primitive streak formation negative regulation of canonical Wnt signaling pathway cochlea morphogenesis melanocyte proliferation presynapse assembly postsynapse assembly postsynaptic modulation of chemical synaptic transmission regulation of postsynapse organization regulation of postsynaptic cytosolic calcium ion concentration meiotic nuclear division positive regulation of neuron projection arborization positive regulation of protein kinase C activity positive regulation of hematopoietic stem cell proliferation positive regulation of protein localization to synapse excitatory synapse assembly inhibitory synapse assembly negative regulation of cell proliferation in midbrain midbrain dopaminergic neuron differentiation positive regulation of cell-cell adhesion mediated by cadherin positive regulation of non-canonical Wnt signaling pathway positive regulation of stem cell proliferation positive regulation of apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline