This antibody can both recognize WNT7A and WNT7B.
Wnt proteins constitute a family of secreted glycoproteins that activate signal transduction pathways to mediate tissue homeostasis, cell fate, cell proliferation, and self-renewal. The canonical signaling pathway is initiated upon Wnt binding to a Frizzled family receptor and an LRP5/LRP6 coreceptor, which leads to activation of the beta-catenin cascade. Non-canonical Wnt signals are activated by binding to a Frizzled family receptor and ROR2 coreceptor. Extensive research has shown that deregulation of these pathways or its components can lead to cancer. Human Wnt-1 is encoded as a 370 amino acid precursor that, upon secretion, is cleaved into a 343 amino acid (42 kDa) mature form. At the amino acid level, mature human Wnt-1 is approximately 99% identical to mature mouse Wnt-1. Depending on the tissue, Wnt-1 can activate the canonical and non-canonical signaling pathways. Studies have demonstrated its involvement in a variety of functions, including embryonic development and carcinogenesis. Wnt-1 signaling can also induce the phosphorylation and subsequent degradation of c-Myb.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: postaxial hemimelia; Protein Wnt-7a; Protein Wnt-7b; proto-oncogene Wnt7a protein; wingless-related MMTV integration site 7A; wingless-related MMTV integration site 7B; wingless-type MMTV integration site 7A; wingless-type MMTV integration site family member 7A; wingless-type MMTV integration site family member 7B; wingless-type MMTV integration site family, member 7A; wingless-type MMTV integration site family, member 7B
Gene Aliases: AI849442; px; tw; Wnt-7a; Wnt-7b; WNT7A; WNT7B
UniProt ID: (Human) O00755, (Human) P56706, (Mouse) P24383, (Mouse) P28047
Entrez Gene ID: (Human) 7476, (Human) 7477, (Rat) 114850, (Mouse) 22421, (Rat) 315196, (Mouse) 22422
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support