Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: PLEVFRRFVW NFFRLENEHL NNCGEFRAVR DISVAPLNAD DQTLLEQMMD QDDGVRNRQK NRSWKYNQSI SLRRPRLASQ SKARDTKVLI EDTDDEAN
Xenotropic and polytropic retrovirus receptor (XPR1) is an Orphan-U GPCR with an unknown ligand. ESTs have been isolated from brain, breast, kidney, pancreas, placenta, prostate, skeletal muscle, testis, and tonsil libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: phosphate exporter SLC53A1; protein SYG1 homolog; RP11-46A10.1; solute carrier family 53 (phosphate exporter), member 1; X-receptor; xenotropic and polytropic murine leukemia virus receptor X3
Gene Aliases: IBGC6; SLC53A1; SYG1; X3
UniProt ID: (Human) Q9UBH6
Entrez Gene ID: (Human) 9213
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support