Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Between 1650 and C-term |
Conjugate |
Unconjugated |
Form |
liquid |
Concentration |
40 µg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
TBS with 0.1% BSA |
Contains |
0.09% sodium azide |
Storage conditions |
4° C |
The recommended shelf life for this product is 1 year from date of receipt.
SEC24C is a member of the SEC24 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec24p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The product of this gene may play a role in shaping the vesicle, as well as in cargo selection and concentration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-Sep; 5'-3' exoribonuclease 1; DKFZp434P0721; DKFZp686B22225; DKFZp686F19113; FLJ41903; protein Dhm2; Strand-exchange protein 1 homolog
Gene Aliases: Dhm2; exo; mXrn1; SEP1; XRN1
UniProt ID: (Human) Q68D88
Entrez Gene ID: (Human) 54464, (Mouse) 24127
Molecular Function:
exoribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support