YIF1B belongs to the YIF1 family. It is a multi-pass membrane protein. The functions of YIF1B remain unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein YIF1B; Yip1 interacting factor homolog B; YIP1-interacting factor homolog B
Gene Aliases: FinGER8; PP4519; UNQ3073/PRO9905; YIF1B
UniProt ID: (Human) Q5BJH7
Entrez Gene ID: (Human) 90522
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support