Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: VTMWSSDEKK DIDHETVVEE QIIGENSPPD YSEYMTGKKL PPGGIPGIDL SDPKQLAEFA RMKPRKIKED DAPRTIACPH KGCTKMFRDN SAMRKHLHTH
YY1 is a ubiquitously distributed transcription factor belonging to the GLI-Kruppel class of zinc finger proteins. The protein is involved in repressing and activating a diverse number of promoters. YY1 may direct histone deacetylases and histone acetyltransferases to a promoter in order to activate or repress the promoter, thus implicating histone modification in the function of YY1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Delta transcription factor; INO80 complex subunit S; NF-E1; Transcriptional repressor protein YY1; Yin and yang 1; Yin and Yang 1 protein; YY-1
Gene Aliases: DELTA; INO80S; NF-E1; UCRBP; YIN-YANG-1; YY1
UniProt ID: (Human) P25490
Entrez Gene ID: (Human) 7528
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support