Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AFSQYFRSLF QNSSSQKNDV FHLDVKNVSG IGQILDFMYT SHLDLNQDNI QVMLDTAQCL QVQNVLSLCH TFLKSATVVQ PPGMPCNSTL SLQSTLTPDA TCVISENYP
Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 79%.
Transcription factor. Inhibits cell proliferation by activating either CDKN1A/p21 transcription or RB1 transcription. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and BTB domain-containing protein 49; Zinc finger protein 509
Gene Aliases: ZBTB49; Zfp509; ZNF509
UniProt ID: (Human) Q8TB80
Entrez Gene ID: (Human) 166793, (Rat) 305428
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support