Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MEHLGDSKLQ RDVSLDSKYR ETCKRDSKAE KQQAHSTGER RHRCNECGKS FTKSSVLIEH QRIHTGEKPY ECEECGKAFS RRSSLNEHRR SHTGEKPYQC KECGKAFSAS NGLTRHRRIH TGEKPYECKV CGKAFLLSSC LVQHQRIHTG EKRYQCRECG KAFIQNTGLF QHLRVHTGEK PYQCSQCSKL FSKRTLLRKH QKIHTGERP
ZNF323 is a member of the subfamily of C2H2 Kruppel-like zinc finger transcription factors that have a SCAN box domain (Pi et al., 2002 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ874C20.2; Zinc finger and SCAN domain-containing protein 31; Zinc finger protein 323
Gene Aliases: ZNF20-Lp; ZNF310P; ZNF323; ZSCAN31
UniProt ID: (Human) Q96LW9
Entrez Gene ID: (Human) 64288
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support