Search
Search
NeoBiotechnologies
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody is stable for 24 months.
Positive Control: HeLa or 293T cells.Human testis, spleen or colon. Cellular Localization: Nucleus. Cytoplasm.
Specificity Comments: Zinc finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc finger proteins contain a Kr ppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the Kr ppel C2H2-type zinc finger protein family, ZNF358 (zinc finger protein 358) is a 481 amino acid nuclear protein that contains nine C2H2-type zinc fingers through which it is thought to be involved in DNA-binding and transcriptional regulation.
ZNF358, a gene located on chromosome 19, encodes a protein involved in embryogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10390; Zinc finger protein 358
Gene Aliases: ZFEND; ZNF358
UniProt ID: (Human) Q9NW07
Entrez Gene ID: (Human) 140467
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support