Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1-5 µg/mL | - |
Immunocytochemistry (ICC/IF) |
10 µg/mL | - |
ELISA (ELISA) |
10 µg/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG1, kappa |
Class |
Monoclonal |
Type |
Antibody |
Clone |
1F8 |
Immunogen |
ZNF394 (AAH25241, 1 a.a. approximately 561 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
See Label |
Purification |
Affinity chromatography |
Storage buffer |
PBS, pH 7.4 |
Contains |
no preservative |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
Sequence of this protein is as follows: MNSSLTAQRR GSDAELGPWV MAARSKDAAP SQRDGLLPVK VEEDSPGSWE PNYPAASPDP ETSRLHFRQL RYQEVAGPEE ALSRLRELCR RWLRPELLSK EQILELLVLE QFLTILPEEL QAWVREHCPE SGEEAVAVVR ALQRALDGTS SQGMVTFEDT AVSLTWEEWE RLDPARRDFC RESAQKDSGS TVPPSLESRV ENKELIPMQQ ILEEAEPQGQ LQEAFQGKRP LFSKCGSTHE DRVEKQSGDP LPLKLENSPE AEGLNSISDV NKNGSIEGED SKNNELQNSA RCSNLVLCQH IPKAERPTDS EEHGNKCKQS FHMVTWHVLK PHKSDSGDSF HHSSLFETQR QLHEERPYKC GNCGKSFKQR SDLFRHQRIH TGEKPYGCQE CGKSFSQSAA LTKHQRTHTG EKPYTCLKCG ERFRQNSHLN RHQSTHSRDK HFKCEECGET CHISNLFRHQ RLHKGERPYK CEECEKSFKQ RSDLFKHHRI HTGEKPYGCS VCGKRFNQSA TLIKHQRIHT GEKPYKCLEC GERFRQSTHL IRHQRIHQNK VLSAGRGGSR L
ZNF394 belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SNAP19; truncated zinc finger protein isoform; Zinc finger protein 394; zinc finger protein 99; Zinc finger protein with KRAB and SCAN domains 14
Gene Aliases: ZKSCAN14; ZNF394; ZSCAN46
UniProt ID: (Human) Q53GI3
Entrez Gene ID: (Human) 84124
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support