Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: GNSERRDILM DKLAPLGRPY ESLTVQLHPK KTQLEQEAGK PQRNGDKTRT KNEELFQKED MPKDKEFLGE INDRLNKDTP QHPKSKDIIE NEGRSEWQQ
ZNF435 contains 1 SCAN box domain and 4 C2H2-type zinc fingers. It belongs to the kruppel C2H2-type zinc finger protein family and may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and SCAN domain-containing protein 16; Zinc finger protein 392; Zinc finger protein 435
Gene Aliases: dJ265C24.3; ZNF392; ZNF435; ZSCAN16
UniProt ID: (Human) Q9H4T2
Entrez Gene ID: (Human) 80345
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support