Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: IPSPDADLAT SGTHTVTMVS ADGTQTQPVT IITSGAVVAE DSSVASLRHQ QVALLATANG THIAVQLEEQ QTLEEAINVA TAAMQQGAVT LETTVSESGC
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: unnamed protein product; Zinc finger protein 523; Zinc finger protein 76; zinc finger protein 76 (expressed in testis)
Gene Aliases: D6S229E; Zfp523; ZNF523; ZNF76
UniProt ID: (Human) P36508
Entrez Gene ID: (Human) 7629
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support