Search
Search
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
ZSCAN21 belongs to the krueppel C2H2-type zinc-finger protein family and is a strong transcriptional activator.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Renal carcinoma antigen NY-REN-21; Zfp-38; Zinc finger and SCAN domain-containing protein 21; zinc finger proliferation 1; zinc finger protein 38 (KOX 25); Zinc finger protein 38 homolog; zinc finger protein NY-REN-21 antigen
Gene Aliases: NY-REN-21; ZFP38; Zipro1; ZNF38; ZSCAN21
UniProt ID: (Human) Q9Y5A6
Entrez Gene ID: (Human) 7589, (Rat) 304342
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support